Learn More
Cayman Chemical Goat ntI-MouseH L HRP 1mg
SDP
Supplier:  Cayman Chemical 167381 MG
Encompass_Preferred
Goat anti-Mouse IgG, H+L conjugated to HRP Antibody: Goat polyclonal anti-Mouse IgG, H+L Enzyme: Horseradish Peroxidase Uses: ELISA and Western blot detection. Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000.
Catalog No. 50-296-5121