Learn More
Cayman Chemical Goat ntIHumn IgG EuW1024 50ug
SDP
Supplier:  Cayman Chemical 1657150 UG
Encompass_Preferred
Goat Anti-Human IgG, secondary antibody, conjugated to Europium 1024, 20μg Concentration: 1 mg/ml Antibody: Goat poyclonal anti-Human IgG Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications Form: Conjugate supplied as a liquid in 50 mn Tris (pH 7.8) + 0.9% NaCl + 0.05% NaN3.
Catalog No. 50-296-7279