Learn More
Cayman Chemical Goat ntIlama IgG SureLIght 1ea
SDP
Supplier:  Cayman Chemical 134151 EA
Encompass_Preferred
Goat anti-Llama IgG conjugated to SureLight™ R-PE Antibody: Goat polyclonal anti-Llama IgG Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-295-4792