Learn More
Cayman Chemical Goat ntIRat IgG FcSpec Sur
SDP
Supplier:  Cayman Chemical 16654100 UG
Encompass_Preferred
Goat anti-Rat IgG, Fc-specific conjugated to SureLight™ R-PE Antibody: Goat polyclonal anti-Rat IgG, Fc-specific Dye: SureLight™ R-phycoerythrin (R-PE) Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-296-6606