Learn More
Medchemexpress LLC Goflikicept 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP996451MG
Encompass_Preferred
Goflikicept (RPH 104) is a fusion protein that selectively binds and inactivates both circulating IL-1 and IL-1 Goflikicept has the potential for the research of ST-segment elevation myocardial infarction (STEMI)[1]
Catalog No. 50-003-22604