Learn More
Essendant GOGGLESSAFETYCLR
Supplier:  Essendant CRW2220BX
Encompass_Preferred
Provides a wide unobstructed field of vision and can be worn with most half-mask respirators Transparent PVC body permits a comfortable fit over glasses Protects against impact splash Economy Goggles Perforated ventilation Material(s)- PVC For Use With- Over Glasses Lens Tint- Clear Side Protection- Yes
Catalog No. 50-000-19742