Learn More
Essendant GOGGLESWLDCATSFTYCL AF
Supplier:  Essendant ANS20525
Encompass_Preferred
Heat-resistant goggles will not melt drip or ignite at 350 degrees for five minutes Dual anti-fog lenses provide clear vision Lightweight and comfortable enough for any work environment Adjustable side vents increase air flow Flame-retardant pivoting headband holds goggles securely and allows for comfortable placement Cushioned eyecups provide comfort for extended wear Adjustability Features- Strap Material(s)- Polycarbonate Antifog- Yes Coated Lenses- Yes
Catalog No. 50-000-20369