Learn More
Enterprise Technology Solutions Gojo Hand Sanitizer,Gel,f/CS4 Manual,VF Plus,1200ml,4/CT,CL
SDP
Supplier:  Enterprise Technology Solutions GOJ519904
Encompass_Preferred
With broad-spectrum activity, PURELL VF Plus Hand Sanitizer Refills are designed for PURELL CS4 Push-Style Hand Sanitizer Dispensers for professional environments. This sanitizer kills 99.99 percent of germs, including the most common foodborne, bacterial pathogens. It helps reduce germs on hands in high-traffic, high-touch areas, including restaurants and cruise ships. Quick-drying formula feels as clean as PURELL Advanced Hand Sanitizer. It meets Food Code Hand Sanitizers criteria. Hand sanitizer is fragrance-free and dye-free. It is compatible with nitrile, vinyl and latex gloves. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill is easily recycled.
Catalog No. 50-008-11076