Learn More
Medchemexpress LLC Gol-NTR | 2968461-58-5 | C24H16F3N3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15153750MG
Encompass_Preferred
Gol-NTR is a Golgi-targetable probe characterized by high selectivity and sensitivity. It is activated by Nitroreductase (NTR) and is capable of visualizing acute lung injury (ALI) and its repair function.
  • High selectivity and sensitivity
  • Activated by Nitroreductase (NTR)
  • Capable of visualizing acute lung injury (ALI) and its repair function
  • Low detection limit of 54.8 ng/mL
  • Can be utilized for monitoring and assessing the response to sepsis-induced ALI
Catalog No. 50-003-73954