Learn More
Abcam Gold Conjugation Kit (10 nm, 20 OD), 1 x 25UG
Supplier:  Abcam AB2018081X25UG
Encompass_Preferred
10nm Gold conjugation / labeling in < 4 hrs with 30 secs hands-on time using Gold Conjugation Kit (10 nm, 20 OD) ab201808. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9114