Learn More
Strem, An Ascensus Company CAS# 7440-57-5. 25 x 25mm. Gold foil (99.9%). MFCD00003436
SDP
Supplier:  Strem, An Ascensus Company 79005025X25MM
Encompass
CAS# 7440-57-5. 25 x 25mm. Gold foil (99.9%). MFCD00003436. Molecular Weight: 197.20. Molecular Formula: Au. Color/form: 0.01mm thick (~0.12g/25mm x 25mm) Strem# 79-0050. http://www.strem.com/catalog/v/79-0050/
Catalog No. 50-901-13341