Learn More
Sigma Aldrich Fine Chemicals Biosciences Gold foil, thickness 0.007 mm, size 50 x 50 mm, purity 99.9% | 7440-57-5 | MFCD00003436 | 1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GF806323021EA
Encompass
Gold foil, thickness 0.007 mm, size 50 x 50 mm, purity 99.9% | Purity: 99.9% | Mol Wt: 196.97 | 7440-57-5 | MFCD00003436 | 1EA
Catalog No. 50-185-1384