Learn More
Strem, An Ascensus Company Gold(III) chloride 3N 5g
SDP
Supplier:  Strem, An Ascensus Company 9379075G
Encompass
CAS# 13453-07-1. 5g. Gold(III) chloride, 99% (99.9%-Au). MFCD00014172. Molecular Weight: 303.33. Molecular Formula: AuCl3. Color/form: red-brown solid Strem# 93-7907. http://www.strem.com/catalog/v/93-7907/
Catalog No. 50-901-16392