Learn More
Nanocomposix NanoXact Gold Nanorods - Bare (Citrate) - Peak @ 980 nm · 80 nm × 15 nm, 1 OD in water, 25 mL
SDP
Supplier:  Nanocomposix GRCN98025M
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.980 nm peak resonance (SPR) gold nanorods combine strong plasmonic optical properties with unique anisotropic morphology. These nanoparticles are useful in a broad range of applications from immunodiagnostics, biosensors, sensing, photothermal therapy, SERS, optical filters, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC1674772