Learn More
Medchemexpress LLC Gold sodium thiomalate | 12244-57-4 | MFCD00064304 | 98.0% | 390.08 g/mol | C4H6O4S·Au·xNa | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1063815MG
Encompass_Preferred
Aurothiomalate sodium is a gold-containing research reagent reported as an anti-rheumatic agent that functions as a potent protein kinase C iota (PKCι) signaling inhibitor and a thioredoxin reductase (TrxR) inhibitor. It is supplied as a solid for biochemical and cellular research applications.
  • Potent PKCι and thioredoxin reductase (TrxR) inhibitor.
  • Reported anti-rheumatic agent with demonstrated anti-tumor activity.
  • Supplied as a solid suitable for biochemical and cellular studies.
  • High purity (98.0%) for research applications.
  • Stored sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-43340