Learn More
Med Vet International Golden, A5, Two-Speed, Clipper
Supplier:  Med Vet International CLIPPERGOLDA5
Encompass_Preferred
These versatile clippers with an insulated face plate for cooler running operation will handle everything from general purpose grooming to precision clipping, on any coat type. Smooth, quiet, reliable, and cool-running, with a powerful Universal motor. -Two Speed (up to 2700 strokes per minute) -Wider sweep for faster cutting-Compatible with A5 Cryogen-X™ & Elite Cryogen-X™ or Take-Down-Quick™ wide blades-Detachable blade system makes for quick, easy blade changes and increased versatility-12' insulated cord Includes: Cryogen-X™ AgION #10 detachable blade Blade guard Blade oil Clipper grease Cleaning brush Extra brush and spring assembly
Catalog No. 50-283-2326