Learn More
Perkin Elmer US LLC Golden Gate ATR - Single Reflection
Supplier:  Perkin Elmer US LLC L1272407
Encompass_Preferred
The robust design allows this ATR accessory to be used to achieve high-quality spectral data in the analysis of a broad range of materials including those that are abrasive, hard, reactive, or corrosive. This ATR can be ordered with an optional set of ZnSe or KRS-5 lenses. This accessory is compatible with the Spectrum 3/Frontier spectrometer.
Catalog No. 50-211-3283
Only null left
Add to Cart
Request a Quote