Learn More
Cayman Chemical Golgcde A 5mg
SDP
Supplier:  Cayman Chemical 184305 MG
Encompass_Preferred
A reversible inhibitor of the cis-Golgi ArfGEF, GBF1 (IC50 = 3.3 µM); induces rapid dissociation of COPI vesicle coat protein from Golgi membranes and disassembly of the Golgi and trans-Golgi network, preventing bidirectional transport of endocytic cargo
Catalog No. 50-296-6422