Learn More
Medchemexpress LLC Golgicide A-1 | 1394285-49-4 | 99.9% | 284.3 | C17H14F2N2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100540C25MG
Encompass_Preferred
Golgicide A-1 is the cis-diastereomer of Golgicide A and a research-grade small molecule reported to weakly inhibit mosquito reproduction. It displays minimal to no cytotoxicity in Aedes aegypti and Anopheles stephensi larvae assays and is supplied as a solid for laboratory research applications.
  • Cis-diastereomer with distinct isomeric profile
  • Weak inhibitor of mosquito reproduction
  • Minimal to no cytotoxicity in mosquito larvae assays
  • Provided as a powder suitable for laboratory storage
  • Recommended storage conditions and stability data available
Catalog No. 50-002-14482