Learn More
Medchemexpress LLC Golgicide A-2 | 1394285-50-7 | 99.5% | 284.30 g·mol⁻1 | C17H14F2N2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100540B50MG
Encompass_Preferred
Golgicide A-2 is a Golgicide A derivative and the most active enantiomer, supplied for research use to study Golgi apparatus disruption, antiviral activity, and selective insecticidal effects. CAS 1394285-50-7; molecular formula C17H14F2N2; molecular weight 284.30 g·mol⁻1.
  • Most active enantiomer of Golgicide A used in mechanistic studies.
  • Targets Golgi-associated pathways and ArfGEF GBF1-related processes.
  • Reported activity in antiviral and selective insecticidal assays.
  • High purity suitable for biochemical and cell-based assays.
  • Available in milligram-scale quantities for laboratory research.
Catalog No. 50-002-14945