Learn More
Medchemexpress LLC Golgicide A-2 | 1394285-50-7 | 99.5% | 284.30 g/mol | C17H14F2N2 | 1 ML

Supplier: Medchemexpress LLC HY100540B10MM 1ML
Golgicide A-2 is a synthetic enantiomeric derivative of Golgicide A that selectively inhibits cis-Golgi ADP-ribosylation factor guanine nucleotide exchange factors (ArfGEFs). It is used in research on Golgi function, viral replication, and insecticidal activity. The compound is supplied as a ready-to-use solution for laboratory experiments.
- 10 mM in DMSO, 1 mL vial.
- Selective inhibitor of cis-Golgi Arf guanine nucleotide exchange factors (ArfGEFs).
- High purity (99.5%).
- Molecular formula C17H14F2N2 and molecular weight 284.30 g/mol.
- CAS number 1394285-50-7.
- Suitable for studies of viral replication, Golgi function, and insecticidal effects.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.