Learn More
Medchemexpress LLC Golotimod (hydrochloride) | 1029401-59-9 | 99.79% | 369.80 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14743B10MM/1ML
Encompass_Preferred
Golotimod hydrochloride (SCV 07 hydrochloride) is an immunomodulating peptide with antimicrobial activity. It significantly increases the efficacy of antituberculosis therapy, stimulates thymic and splenic cell proliferation, and improves macrophage function. It also inhibits STAT3 signaling and modulates the duration and severity of oral mucositis, showing potential as a therapeutic for recurrent genital herpes simplex virus type 2 (HSV-2).
  • Immunomodulating peptide with antimicrobial activity
  • Enhances antituberculosis therapy efficacy
  • Stimulates thymic and splenic cell proliferation
  • Improves macrophage function
  • Inhibits STAT3 signaling
  • Modulates duration and severity of oral mucositis
  • Potential therapeutic for recurrent genital herpes simplex virus type 2 (HSV-2)
Catalog No. 50-003-60351