Learn More
Medchemexpress LLC Golvatinib | 928037-13-2 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13068
Encompass_Preferred
Golvatinib (E-7050) is a potent dual inhibitor of both c-Met and VEGFR2 kinases with IC50s of 14 and 16 nM, respectively. It effectively inhibits phosphorylation of c-Met and VEGFR-2, repressing the growth of c-Met amplified tumor cells and endothelial cells.
  • Inhibits phosphorylation of c-Met and VEGFR-2
  • Represses growth of c-Met amplified tumor cells and endothelial cells
  • Circumvents resistance to EGFR-TKIs by blocking the Met/Gab1/PI3K/Akt pathway
  • Prevents emergence of gefitinib-resistant cells
  • Shows inhibition of c-Met and VEGFR-2 phosphorylation in tumors
  • Strongly inhibits tumor growth and angiogenesis in xenograft models
Catalog No. 50-003-42628