Learn More
Apexbio Technology LLC Golvatinib (E7050), 10mg. Cas: 928037-13-2 MFCD: MFCD22124462. C-Met/VEGFR-2 inhibitor
SDP
Supplier:  Apexbio Technology LLC B143910
Encompass_Preferred
Golvatinib is an inhibitor of c-Met and VEGFR-2 tyrosine kinase with IC50 values of 14nM and 16nM, respectively [1].The small molecule compound, golvatinib, is a dual inhibitor of both c-Met and VEGFR-2. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5238
 Restricted Item