Learn More
Medchemexpress LLC Gomisin H | 66056-20-0 | 1 MG

Supplier: Medchemexpress LLC HYN22461MG
Gomisin H is a dibenzocyclooctadiene lignan isolated from the fruits of *Schizandra chinensis* BAILL.
- Purity 99.0%
- Molecular weight 418.48
- Molecular formula C23H30O7
- Appearance as white to off-white solid
- Soluble in DMSO at ≥ 50 mg/mL
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.