Learn More
Medchemexpress LLC Gomisin H | 66056-20-0 | 5 MG

Supplier: Medchemexpress LLC HYN22465MG
Gomisin H is a dibenzocyclooctadiene lignan isolated from the fruits of *Schizandra chinensis* BAILL.
- Dibenzocyclooctadiene lignan
- Molecular weight: 418.48
- Formula: C23H30O7
- Appearance: Solid
- Color: White to off-white
- Soluble in DMSO: ≥ 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.