Learn More
ATCC Gonapodya polymorpha Thaxter - ATCC MYA-4798, frozen ampoule culture (JEL 455), BSL 1
Supplier: ATCC MYA4798
Gonapodya polymorpha is supplied as a frozen ampoule containing viable cells suspended in cryoprotectant. The genome-sequenced JEL 455 strain was isolated from a vernal pool in Bethel, Maine, and is intended for laboratory research requiring specialized handling; viability may take up to 14-18 days to observe.
- Provided as a frozen ampoule in cryoprotectant.
- Genome-sequenced strain (JEL 455).
- Isolation source: vernal pool; Bethel, Maine, USA.
- Recommended incubation: 20-25°C, aerobic.
- Growth medium: mPmTG specialized fungal medium.
- Storage: -80°C or colder; do not store at -20°C.
- Handling: thaw in a 25°C to -30°C water bath; avoid agitation; aseptically transfer ≥50 μL or 2-3 agar cubes.
- Growth characteristics: slow growth; inspect for viability after 14-18 days.
- Biosafety level: BSL 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.