Learn More
Medchemexpress LLC GoSlo-SR-5-69 | 1363419-31-1 | N/A | 99.3% | 470.47 g·mol-1 | C24H19N2NaO5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1310125MG
Encompass_Preferred
GoSlo-SR-5-69 is a small-molecule activator of large-conductance, calcium-activated potassium (BK) channels. It potentiates BK channel opening in electrophysiological and cellular assays (reported EC50 = 251 nM) and is used to probe BK-mediated currents and physiological responses. Supplied as a sodium sulfonate salt with high analytical purity.
  • Potent BK channel activation (EC50 = 251 nM).
  • High purity, tested by RP-HPLC (>99%).
  • Supplied as a sodium sulfonate salt.
  • Soluble in DMSO for assay preparation.
  • Suitable for electrophysiology and ion-channel pharmacology studies.
  • Provided in small research pack sizes for laboratory use.
Catalog No. 50-002-64919