Learn More
Medchemexpress LLC Gosogliptin | 869490-23-3 | 99.8% | 366.41 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY10287
Encompass_Preferred
Gosogliptin is a potent and selective inhibitor of dipeptidyl peptidase-IV (DPP-IV). It is an orally active, selective, and competitive inhibitor of DPP-IV, the enzyme primarily responsible for the degradation of incretin peptides GLP-1 and glucose-dependent insulinotropic polypeptide. In various nonclinical models, Gosogliptin stimulates insulin secretion and improves glucose tolerance.
  • Demonstrates more than 200-fold selectivity over other DPP family members and related serine proteases
  • Shows rapid and reversible inhibition of plasma DPP-4 activity
  • Stimulates insulin secretion and improves glucose tolerance
  • For research use only
Catalog No. 50-003-65742