Learn More
Medchemexpress LLC Gossypolone | 4547-72-2 | 96.3% | 1 MG

Supplier: Medchemexpress LLC HY1197341MG
Gossypolone is a small-molecule inhibitor of the RNA-binding protein Musashi-1 used for preclinical and research applications, particularly in cancer research. Supplied as a light yellow to brown solid with stated purity of about 96.3%, it is intended for laboratory research use only and should be stored under nitrogen at 4°C; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
- Small-molecule inhibitor of Musashi-1 (MSI1)
- CAS number 4547-72-2
- Molecular formula C30H26O10, molecular weight 546.52 g/mol
- Purity 96.3%
- Appearance: light yellow to brown solid
- Intended for laboratory research use only
- Recommended storage: under nitrogen at 4°C; in solution, -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.