Learn More
Medchemexpress LLC HY-122723 5mg Medchemexpress, GOT1 inhibitor-1 CAS:732973-87-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1227235MG
Encompass_Preferred
Medchemexpress, HY-122723 5mg GOT1 inhibitor-1 CAS:732973-87-4 GOT1 inhibitor-1 (compound 2c), a tryptamine-based derivative, acts as a novel, potent and non-covalent inhibitor of glutamate oxaloacetate transaminase 1 (GOT1) with an IC50 of 8.2 uM. GOT1 inhibitor-1 can be used for the research of pancreatic ductal adenocarcinoma (PDAC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0993