Learn More
Medchemexpress LLC Got1 inhibitor-1 | 732973-87-4 | 98.6% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12272325MG
Encompass_Preferred
GOT1 inhibitor-1 is a tryptamine-based, non-covalent inhibitor of glutamate oxaloacetate transaminase 1 (GOT1) with a reported IC50 of 8.2 μM. It is supplied as a solid for biochemical and cellular research into metabolism and reactive oxygen species balance, including pancreatic ductal adenocarcinoma studies.
  • Non-covalent inhibitor of GOT1 with reported IC50 8.2 μM.
  • High analytical purity suitable for research use.
  • Available in multiple solid sizes, including 25 mg.
  • Stable when stored as powder at -20°C for long-term storage.
  • Useful for biochemical assays and cellular metabolism studies.
Catalog No. 50-002-30935