Learn More
Abcam Gouml 6983, PKC inhibitor, 1MG
Supplier:  Abcam AB1444141MG
Encompass_Preferred
MW 442.5 Da, Purity >98%. Potent, broad spectrum PKC inhibitor (IC50 values are 7, 7, 6, 10, 60 and 20000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ and PKCμ respectively). Shows cardioprotective and anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5775