Learn More
Medchemexpress LLC GP(33-41) TFA | 99.6% | 1130.19 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0323A10MG
Encompass_Preferred
GP(33-41) TFA is a 9-amino acid-long peptide that represents the optimal sequence of the GP1 epitope of the lymphocytic choriomeningitis virus. It can upregulate H-2Db molecules at the RMA-S (Db Kb) cell surface with an SC50 of 344 nM. In vitro, it sensitizes MC57 and T2-Db cells to lysis with ED50s of 0.9±0.6 and 2.5±0.7 nM, respectively. The P14 TCR recognizes this peptide when presented in the context of H-2D.
  • Optimal sequence of the GP1 epitope of lymphocytic choriomeningitis virus
  • Upregulates H-2Db molecules at the RMA-S (Db Kb) cell surface
  • Sensitizes MC57 and T2-Db cells to lysis with low nanomolar ED50 values
  • Recognized by P14 TCR in the context of H-2D
Catalog No. 50-004-23779