Learn More
Cayman Chemical GP-AMChydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 3445810 MG
Encompass_Preferred
A fluorogenic substrate for DPP-4; AMC is released upon enzymatic cleavage by DPP-4 and its fluorescence can be used to quantify DPP-4 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-295-0334