Learn More
MSC AbilityOne General Purpose Detergent, Liquid, MIL-D-16791G Type I, 5 Gallon
Supplier:  MSC 80423171
Encompass
The AbilityOne General Purpose Detergent is a liquid, nonionic cleaning agent designed to meet MIL-D-16791G, Type I specifications. Supplied in a 5 gallon pail, it offers effective cleaning for various applications.
  • Liquid form
  • Nonionic formula
  • Meets MIL-D-16791G, Type I specification
  • 5 gallon pail container
  • Neutral pH of 7
  • Highly soluble in water (>10%)
  • Chemically stable
  • Manufactured in the United States
Catalog No. 50-283-9075