Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC GP+E-86 (CRL-3620) Mouse embryo fibroblast cell line, retroviral packaging and transfection host

Supplier:  ATCC CRL3620

Encompass

A mouse embryo fibroblast cell line engineered to package ecotropic retroviral genomes and serve as a transfection host for production of retroviral vectors for delivery of foreign genes into susceptible eukaryotic cells. Derived from NIH 3T3 cells via electroporation of plasmids encoding gag/pol and env, it is commonly used for retroviral vector production and transfection workflows.

  • Suitable for packaging psi-containing nucleic acids into ecotropic retrovirus genomes.
  • Adherent growth with fibroblast morphology.
  • Recommended culture at 37°C in 95% air, 5% CO2.
  • Complete medium: DMEM supplemented with hypoxanthine (0.015 mg/mL), xanthine (0.25 mg/mL), mycophenolic acid (0.025 mg/mL), and 90% calf serum.
  • Subculture ratio typically 1:5 to 1:10; renew medium twice weekly.
  • Store cryogenically in vapor phase of liquid nitrogen; cryopreserve in medium with 5% DMSO.

Catalog No. NC3845706


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.