Learn More
Medchemexpress LLC Recombinant human gp130/IL6ST protein | >95.0% | 50UG

Supplier: Medchemexpress LLC HYP70131950UG
Recombinant human gp130/IL6ST extracellular domain expressed in HEK293 cells and supplied as a lyophilized powder. The protein carries a C-terminal His tag and is formulated from PBS, pH 7.4, with 8% trehalose to support stability for biochemical and cell-based studies.
- Recombinant extracellular domain (E23-I618) with C-terminal 8xHis tag.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted molecular weight 78-110 kDa for the glycosylated protein.
- Lyophilized from PBS, pH 7.4, with 8% trehalose for enhanced stability.
- Store at -20°C; reconstituted material stable at 4°C for 1 week or -20°C for longer.
- Produced in HEK293 mammalian cells to preserve native glycosylation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.