Learn More
Santa Cruz Biotechnology gp91-phox Antibody, 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC130543
Encompass
Anti-gp91-phox Antibody (54.1) is recommended for detection of gp91-phox of mouse, rat and human origin by WB, IP, IF and IHC(P); also recommended for detection of ERp57, Mox1, Nox3 and Nox4.
Catalog No. NC1794408