Learn More
Selleck Chemical LLC gp91ds-tat P1213-5MG
SDP
Supplier:  Selleck Chemical LLC P12135MG
Encompass_Preferred
gp91ds-tat a peptide inhibitor for NADPH oxidase assembly is composed of gp91phox sequence linked to the human immunodeficiency virus-tat peptide The tat sequence facilitates the entry of this peptide into cells
Catalog No. 50-000-42671