Learn More
KACTUSBIO INC Human GPC3 Protein-VLP (GPC-HE005) . 1EA=100UG.
Supplier:  KACTUSBIO INC GPCHE005100UG
Encompass
Recombinant Human GPC3 Protein-VLP is expressed from E.coli.It contains Gly510-Asn554.
Catalog No. NC2956820