Learn More
KACTUSBIO INC Human GPC3 (438-554) Protein-VLP (GPC-HM003) . 1EA=100UG.
Supplier:  KACTUSBIO INC GPCHM003100UG
Encompass
Recombinant Human GPC3 (438-554) Protein-VLP is expressed from HEK293.It contains Arg438-Asn554.
Catalog No. NC2956822