Learn More
Abcam GpCp (GMPCP), GDP methylene analog, 25MG
Supplier:  Abcam AB14666325MG
Encompass_Preferred
MW 441.23 g/mol, Purity >95%. GDP methylene analog. Contains methylene group replacing the α and β oxygen. Incorporates into hydrolyzable microtubule subunits. Related GTP analog GMPPCP (ab146660) is also available.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4540