Learn More
Medchemexpress LLC Gphos Pd G6 | 2489525-81-5 | MFCD34602234 | 98.5% | 944.44 g/mol | C47H70BrO4PPdSi | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW590989250MG
Encompass_Preferred
GPhos Pd G6 (GPhos Pd G6 TES) is a palladium-based phosphine reagent used as a drug intermediate in organic synthesis and medicinal chemistry. It is supplied as a solid reagent with documented purity and accompanying batch documentation for quality and safety.
  • High purity (≈98.5%) suitable for synthetic applications.
  • Provided with batch COA and SDS for verification and safe handling.
  • Available in small-scale pack sizes, including 250 MG, for method development and synthesis.
  • Recommended storage: sealed, protected from moisture at 4°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Palladium phosphine structure supports cross-coupling and catalytic transformations.
Catalog No. 50-002-84011