Learn More
Medchemexpress LLC GPLGIAGQ TFA | 99.9% | 825.83 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP2213A10MG
Encompass_Preferred
GPLGIAGQ TFA is a polypeptide that is cleavable by Matrix Metalloproteinase-2 (MMP2). It is utilized as a stimulus-sensitive linker in both liposomal and micellar nanocarriers for MMP2-triggered tumor targeting, and can be used in the synthesis of unique MMP2-targeted photosensitizers for photodynamic therapy (PDT).
  • Cleavable by matrix metalloproteinase-2 (MMP2)
  • Functions as a stimulus-sensitive linker in nanocarriers
  • Facilitates MMP2-triggered tumor targeting
  • Used for synthesizing MMP2-targeted photosensitizers in photodynamic therapy (PDT)
  • Triggers PEG deshielding of liposomal carriers
  • Leads to enhanced cellular internalization
  • For research use only
Catalog No. 50-004-22992