Learn More
Eurofins Discoverx ChemiScreen GPR103/Peptide P518 Receptor Stable Cell Line, 2 vials. (HTS189C)
Supplier:  Eurofins Discoverx HTS189C
Encompass
The ChemiScreen GPR103/Peptide P518 Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. QRFP neuropeptides) based activation of QRFPR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses QRFPR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of QRFPR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6035
Add to Cart
Request a Quote