Learn More
Medchemexpress LLC HY-101492 5mg Medchemexpress, GPR120-IN-1 CAS:1599477-75-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014925MG
Encompass_Preferred
Medchemexpress, HY-101492 5mg GPR120-IN-1 CAS:1599477-75-4 GPR120-IN-1 is a selective Gpr120 agonist with a logEC 50 of −7.62. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2150