Learn More
Medchemexpress LLC GPRP acetate | 157009-81-9 | 98.9% | 485.53 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0074A10MG
Encompass_Preferred
GPRP acetate (Gly-Pro-Arg-Pro acetate) is a fibrin polymerization inhibitor that inhibits the interaction of fibrinogen with the platelet membrane glycoprotein IIb/IIIa complex (GPIIb/IIIa). This product is for research use only.
  • Inhibits fibrin polymerization.
  • Inhibits interaction of fibrinogen with platelet membrane glycoprotein IIb/IIIa complex (GPIIb/IIIa).
  • Increases free thrombin generation in vitro.
  • Alleviates DSS-induced colitis in vivo.
Catalog No. 50-004-25214