Learn More
Medchemexpress LLC GPX4-IN-4 | 2920221-53-8 | 98.9% | 460.93 | 5 MG

Supplier: Medchemexpress LLC HY1499235MG
GPX4-IN-4 (Compound 24) is a potent GPX4 inhibitor primarily used for cancer research.
- Potent GPX4 inhibitor
- Inhibits HT-1080 cell viability
- Inhibits NCI-H1703 cell viability
- Engages kidney GPX4 and induces PD markers in mice
- Molecular formula: C22H21ClN2O5S
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.