Learn More
Medchemexpress LLC GR 103691 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HY10138210MM 1ML
Encompass_Preferred
GR 103691 is a potent selective dopamine D3 receptor antagonist with a Ki value of 0 4 nM GR 103691 shows more than 100-fold selectivity for human dopamine human (h)D3 over hD4 and hD1 sites1]
Catalog No. 50-002-11766